missing translation for 'onlineSavingsMsg'
Learn More

Butyl Nitrite 95.0+%, TCI America™

$98.79 - $786.63

Chemical Identifiers

CAS 544-16-1
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00002058
InChI Key JQJPBYFTQAANLE-UHFFFAOYSA-N
Synonym 1-butyl nitrite, nitrous acid, butyl ester, n-butyl nitrite, nitrous acid, n-butyl ester, nitrous acid butyl ester, n-butylnitrite, butylnitrite, unii-44p8qg0f3t, ccris 838, n-c4h9ono
PubChem CID 10996
IUPAC Name butyl nitrite
SMILES CCCCON=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N037625ML
View Documents
TCI America
N037625ML
25 mL
Each for $98.79
Only null left
 
N0376500ML
View Documents
TCI America
N0376500ML
500 mL
Each for $786.63
Only null left
 
Description

Description

Specifications

Chemical Identifiers

544-16-1
103.121
JQJPBYFTQAANLE-UHFFFAOYSA-N
10996
CCCCON=O
C4H9NO2
MFCD00002058
1-butyl nitrite, nitrous acid, butyl ester, n-butyl nitrite, nitrous acid, n-butyl ester, nitrous acid butyl ester, n-butylnitrite, butylnitrite, unii-44p8qg0f3t, ccris 838, n-c4h9ono
butyl nitrite

Specifications

544-16-1
76°C
MFCD00002058
2351
JQJPBYFTQAANLE-UHFFFAOYSA-N
butyl nitrite
10996
≥95.0% (GC)
Butyl Nitrite
Yellow
C4H9NO2
25 mL
1-butyl nitrite, nitrous acid, butyl ester, n-butyl nitrite, nitrous acid, n-butyl ester, nitrous acid butyl ester, n-butylnitrite, butylnitrite, unii-44p8qg0f3t, ccris 838, n-c4h9ono
CCCCON=O
103.121
103.12
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1565

RTECSNumber : RA0780000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'