missing translation for 'onlineSavingsMsg'
Learn More

Butyl Alcohol, Reagent, ACS, 99.4%, Spectrum™ Chemical

B1185, 71-36-3, C4H10O

$232.53 - $10933.10

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
IUPAC Name butan-1-ol
SMILES CCCCO
View More Specs

Products 9
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18601153
View Documents
Spectrum Chemical Mfg Cor
B1185100ML
100 mL Amber Glass Bottle
Each for $232.53
Only null left
 
18601154
View Documents
Spectrum Chemical Mfg Cor
B1185500MLGL
500 mL Amber Glass Bottle
Each for $267.73
Only null left
 
18601156
View Documents
Spectrum Chemical Mfg Cor
B11851LT
1 L Amber Glass Bottle
Each for $345.43
Only null left
 
18601155
View Documents
Spectrum Chemical Mfg Cor
B1185500MLGLCS6
6 x 500 mL Amber Glass Bottle
Case of 6 Each for $781.50
Only null left
 
18601157
View Documents
Spectrum Chemical Mfg Cor
B11854LTGL
4 L Amber Glass Bottle
Each for $855.97
Only null left
 
18601158
View Documents
Spectrum Chemical Mfg Cor
B11851LTCS6
6 x 1 L Amber Glass Bottle
Case of 6 Each for $1,412.37
Only null left
 
18601159
View Documents
Spectrum Chemical Mfg Cor
B11854LTGLCS4
4 x 4 L Amber Glass Bottle
Case of 4 Each for $2,039.43
Only null left
 
18601160
View Documents
Spectrum Chemical Mfg Cor
B118520LTMT
20 L Steel Can
Each for $1,977.57
Only null left
 
18601161
View Documents
Spectrum Chemical Mfg Cor
B1185200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Butyl Alcohol, Reagent, ACS is more commonly known as butanol. It is also widely used as a solvent in textile and chemical fabrication processes. It can be used as a paint thinner and base for perfumes. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.
Specifications

Chemical Identifiers

71-36-3
74.12
butan-1-ol
C4H10O
LRHPLDYGYMQRHN-UHFFFAOYSA-N
CCCCO

Specifications

71-36-3
100 mL
0.005%
C4H10O
0.1%
CCCCO
74.12
ACS
100%
Amber Glass Bottle
0.0008 meq/g
UN1120
LRHPLDYGYMQRHN-UHFFFAOYSA-N
butan-1-ol
99.4%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'