missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl acetate, CHROMASOLV™ Plus, for HPLC, 99.7%, Honeywell Riedel-de Haën™
CHROMASOLV™ Plus, for HPLC, 99.7%
$341.70 - $1780.53
Chemical Identifiers
CAS | 123-86-4 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00009445 |
InChI Key | DKPFZGUDAPQIHT-UHFFFAOYSA-N |
Synonym | n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle |
PubChem CID | 31272 |
ChEBI | CHEBI:31328 |
IUPAC Name | butyl acetate |
SMILES | CCCCOC(C)=O |
Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Chemical Identifiers
123-86-4 | |
116.16 | |
DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
31272 | |
butyl acetate |
C6H12O2 | |
MFCD00009445 | |
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
CHEBI:31328 | |
CCCCOC(C)=O |
Specifications
123-86-4 | |
100.0000% | |
0.88Â g/mLÂ at 25Â °C(lit.) | |
27°C | |
C6H12O2 | |
MFCD00009445 | |
UN1123 | |
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
CCCCOC(C)=O | |
116.16 | |
CHEBI:31328 | |
>99.65% | |
Butyl acetate, CHROMASOLV™ Plus, for HPLC, 99.7% |
max. 0.01 at 400 nm, max. 0.01 at 320 nm, max. 0.02 at 300 nm, max. 0.04 at 275 nm, max. 0.20 at 260 nm | |
-78°C (literature) | |
126.2°C | |
Glass Bottle | |
CH3COO(CH2)3CH3 | |
1 L | |
1741921 | |
DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
butyl acetate | |
31272 | |
116.16 | |
HPLC |
Safety and Handling
ShelfLife : 900 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Butyl acetates
EINECSNumber : 204-658-1
Recommended Storage : Room Temp