missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Nomex™ IIIA HRC1 Lab Coats
Constructed with flame-resistant Nomex IIIA fabric.
$294.24 - $304.07
Specifications
Product Type | Flame-resistant Lab Coat |
---|---|
Gender | Unisex |
Material | Nomex IIIA, Aramid Fiber |
Color | Royal Blue |
Closure Type | Buttons |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-166-1068
|
Bulwark™
KNL2RBRGS |
Small |
Each for $304.07
|
|
|||||
19-166-1070
|
Bulwark™
KNL2RBRGL |
Large |
Each for $304.04
|
|
|||||
19-166-1072
|
Bulwark™
KNL2RBRGXXL |
2X-Large |
Each for $294.24
|
|
|||||
19-166-1071
|
Bulwark™
KNL2RBRGXL |
X-Large |
Each for $294.24
|
|
|||||
19-295-002
|
Bulwark™
KNL2RBRG3XL |
3X-Large |
Each for $304.07
|
|
|||||
19-166-1069
|
Bulwark™
KNL2RBRGM |
Medium |
Each for $304.04
|
|
|||||
Description
- Arc Rating: ATPV 5.6 calories/cm2
- Button front closure
- Lapel collar
- Left chest pocket
- Hemmed sleeve ends
- Side vent openings
- Royal blue
- Hazard rating class 1
Specifications
Flame-resistant Lab Coat | |
Nomex IIIA, Aramid Fiber | |
Buttons | |
Non-sterile | |
1 | |
KNL2RBRG | |
5.6 Calories/cm2 | |
Chemical, Oil, Gas and Petrochemical Industries, Foundries, Cutting and Welding | |
6 oz. |
Unisex | |
Royal Blue | |
Reusable | |
Flame | |
Lapel | |
Class 1 | |
ASTM 1506, NFPA 2112 | |
Side Vent Openings | |
0.17 kg |