missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ FR Men's Midweight Excel FR™ Comfortouch™ Flame Resistant Deluxe Insulated Bib Overall
For cold jobs, this Midweight Insulated Bib Overall offers unbeatable warmth with CAT 3 AR/FR protection and deluxe features like rugged, double-reinforced knees. Large patch pockets and bonus utility pockets ensure there's a place for all of your tools.
$495.04 - $552.19
Specifications
Closure Type | Hip Zipper with Concealed Snap |
---|---|
Color | Navy |
Material | Cotton, Nylon |
Construction | Cotton, Nylon, Modacrylic lining |
For Use With (Application) | Excellent barrier to dry particles, microorganisms and non-hazardous liquids |
Catalog Number | Mfr. No. | Size | Style | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Style | Price | Qty | ||||
19-080-319
|
Bulwark™ FR
BLC8NVRGS |
Small | Regular |
Each for $552.19
|
|
||||
19-080-321
|
Bulwark™ FR
BLC8NVRGL |
Large | Regular |
Each for $495.04
|
|
||||
Description
- Certifications Level 1: NFPA 2112, NFPA 70E
- Certifications Level 2: Category 3 Protection
- Hazard:
- Primary Closure: Concealed Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Shell: Flame resistant, 7 oz. (235 g/m2) Twill, Water-repellent, Insulation Package: 14 oz. (474 g/m2)
- Blend: Shell: 88% Cotton/12% Nylon, Insulation Package: 50% Modacrylic/50% Cotton
- Closure: Elasticized shoulder straps with non-conductive adjustable hardwear
- Pocket: Five large patch pockets and hip pocket with closure
- Features:
- Reinforced double knees
- Elasticized shoulder straps with non-conductive adjustable hardwear
- Product Collection: Excel FR Comfortouch
- Protection: ATPV Arc Rating: 36
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Designed to be worn over other clothing. Fit over work boots.
- Gender: Male
- Materials: Cotton/Nylon
- Brand: Bulwark™
Specifications
Hip Zipper with Concealed Snap | |
Cotton, Nylon | |
Excellent barrier to dry particles, microorganisms and non-hazardous liquids | |
Elastic |
Navy | |
Cotton, Nylon, Modacrylic lining | |
5 |