missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Reference Standard, pH 7.70 ± 0.01 at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company 157632
Specifications
Buffer, Reference Standard | |
7558-79-4,7778-77-0,7732-18-5 | |
Colorless | |
1X | |
pH Meter and Electrode | |
Liquid | |
pH Buffer |
pH 7.70± 0.01 at 25 °C | |
97.0,0.87,0.16 | |
100.0,0.9,0.17 | |
Calibration | |
7.7 | |
1 L | |
Natural Poly Bottle |