missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Reference Standard, pH 3.56 ± 0.01 at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company 149816
Specifications
Buffer, Reference Standard | |
877-24-7,7647-01-0,7732-18-5 | |
Colorless | |
1X | |
pH Meter and Electrode | |
Liquid | |
pH Buffer | |
Non-sterile |
pH 3.56± 0.01 at 25°C | |
97.0,1.0,0.06 | |
100.0,1.04,0.06 | |
Calibration | |
3.56 | |
500 mL | |
Natural Poly Bottle | |
Reference Standard |