missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Reference Standard, pH 12.45 ± 0.01 at 25°C, Ricca Chemical
$65.69 - $347.63
Specifications
CAS | 7447-40-7,1310-73-2,7732-18-5 |
---|---|
CAS Max % | 0.37 |
Concentration | 1X |
For Use With (Application) | Calibration |
For Use With (Equipment) | pH Meter and Electrode |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
R161800020
|
Ricca Chemical Company
R161800020PK |
20 x 20 mL | Pack |
Each for $347.63
|
|
||||
1618-16
|
Ricca Chemical Company
161816 |
500 mL | Clear PET-G Bottle |
Each for $65.69
|
|
||||
161832
|
Ricca Chemical Company
161832 |
1 L | Natural Poly Bottle |
Each for $73.63
|
|
||||
13-622-601
|
Ricca Chemical Company
16181 |
4 L | Natural Poly Bottle |
Each for $179.34
|
|
||||
Specifications
7447-40-7,1310-73-2,7732-18-5 | |
1X | |
pH Meter and Electrode | |
Liquid |
0.37 | |
Calibration | |
12.45 | |
pH Buffer |