missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company 160032
Specifications
Buffer, Reference Standard | |
497-19-8,144-55-8,1310-73-2,7732-18-5 | |
100.0,0.53,0.41,0.03 | |
Calibration | |
10 | |
1 L | |
Natural Poly Bottle |
pH 10.00 ± 0.01 at 25 deg C | |
97.0,0.51,0.39,0.03 | |
1X | |
pH Meter and Electrode | |
Liquid | |
pH Buffer |