missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Reference Standard, pH 1.00 ± 0.01 at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company 148932
Specifications
Buffer, Reference Standard | |
7647-01-0,7732-18-5 | |
1X | |
pH Meter and Electrode | |
Liquid | |
pH Buffer |
pH 1.00± 0.01 at 25°C | |
Colorless | |
Calibration | |
1 | |
1 L | |
Natural Poly Bottle |