missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 0.95 ± 0.01 at 25°C, Ricca Chemical

$134.88 - $134.88

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 0.95± 0.01 at 25°C
CAS 7647-01-0,7732-18-5
CAS Min % 97.0,1.2
CAS Max % 100.0,1.25
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
148832
View Documents
Ricca Chemical Company
148832
1 L Natural Poly Bottle
Each for $134.88
Only null left
 
Description

Description

Specifications

Specifications

Buffer, Reference Standard
7647-01-0,7732-18-5
100.0,1.25
1X
pH Meter and Electrode
Liquid
pH 0.95± 0.01 at 25°C
97.0,1.2
Colorless
Calibration
0.95
pH Buffer
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'