missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromophenol Blue Indicator, 0.1% (w/v), Ricca Chemical
$50.47 - $231.75
Specifications
Chemical Name or Material | Bromophenol Blue Indicator |
---|---|
CAS | 62625-28-9,7732-18-5 |
CAS Min % | 98.0,0.1 |
CAS Max % | 100.0,0.1 |
Concentration | 0.1% (w/v) |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
1353-4
|
Ricca Chemical Company
13534 |
120 mL | Natural Poly Bottle |
Each for $50.47
|
|
||||
1353-16
|
Ricca Chemical Company
135316 |
500 mL | Natural Poly Bottle |
Each for $133.94
|
|
||||
135332
|
Ricca Chemical Company
135332 |
1 L | Natural Poly Bottle |
Each for $231.75
|
|
||||
Specifications
Bromophenol Blue Indicator | |
98.0,0.1 | |
0.1% (w/v) | |
Indicator | |
Aqueous Solution | |
4 |
62625-28-9,7732-18-5 | |
100.0,0.1 | |
0.1% W/V | |
Natural Poly Bottle | |
Dark Reddish Purple | |
Liquid |