missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ VITLAB™ Wide-Mouth Reagent Bottles with Screw Caps
VITLAB wide mouth polypropylene reagent bottles with polypropylene screw caps have good chemical resistance. Autoclavable at 121 C for 20 minutes.
$292.53 - $453.13
Specifications
Color | Transparent |
---|---|
Material | Polypropylene |
Sterility | Non-sterile |
Closure Included | Yes |
Autoclavable | Autoclavable |
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Price | Quantity | ||||
14-379-353
|
BRANDTECH™ VITLAB™
101689 |
250 mL | GL 45 |
Pack of 20 Each for $453.13
|
|
||||
14-379-354
|
BRANDTECH™ VITLAB™
101789 |
500 mL | GL 45 |
Pack of 10 Each for $292.53
|
|
||||
Description
- Wide mouth for easier access to contents
- Screw caps help protect agaist outside contamination and leakage
Specifications
Transparent | |
Non-sterile | |
Autoclavable | |
Wide | |
Polypropylene |
Polypropylene | |
Yes | |
No | |
Reagent Bottle | |
Screw Cap |