missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ VITLAB™ Reagent Bottles with Screw Caps
VITLAB™ polypropylene reagent bottles with polypropylene screw caps have good chemical resistance. Autoclavable at 121 C for 20 minutes.
Supplier: BRANDTECH™ VITLAB™ 100889
Description
- Good chemical resistance for long-term storage of liquids.
- Screw caps help protect against outside contamination and leakage.
- Autoclavable at 121 C, according to DIN EN 285
Specifications
169.07 oz., 5000mL | |
169.07 oz. | |
7 in. | |
10/Pk. | |
Yes | |
No | |
315 mm | |
12.4 in. | |
GL 45 | |
Screw Cap |
5000 mL | |
Transparent | |
Polypropylene | |
Non-sterile | |
Autoclavable | |
Threaded | |
Reagent Bottle | |
Polypropylene | |
178 mm |