Learn More
BRANDTECH™ BRAND™ Self-adhesive Plate Sealing Film
Securely seals multiwell plates. Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate.
Supplier:  BRANDTECH™ BRAND™ 701365
 View more versions of this product
Catalog No. 13-878-201
Only null left
Explore available promotions
Description
Description
  • Easily apply to plates and remove without the use of expensive equipment.
  • Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
  • Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
    • Polyethylene
    • Polyester
    • Aluminum
    • Rayon
    • Polypropylene
    • Vinyl
Specifications
Specifications
Clear
Rayon
Not Autoclavable
No
No
Pressure Sensitive
Yes
Gas permeable
Sterile
50 Sheets
Cell and tissue culture
Sealing Film
10.1 mil.
-20°C to +80°C
Product Suggestions
Product Suggestions