missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Post Sign Adapter
Quickly locks in place on top of Brady™ Warning Posts to display any rigid sign you choose
Supplier: Brady™ 92135
Description
- Converts a warning post into a sign holder
- Black polyethylene
- 5.08 × 15.24cm (2 × 6 in.)
Specifications
Polyethylene | |
Sign post | |
6 in. | |
Post Sign Adapter |
1 Ea. | |
2 in. | |
CAUTION Step Up | |
Black |