missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Danger: Nitrogen Signs
For Identifying equipment, rooms and materials that contain or are contaminated with hazardous agents
$20.68 - $35.53
Specifications
Certifications/Compliance | ANSI Z535, OSHA 1910.145 |
---|---|
Quantity | 1 Ea. |
Mounting Type | Surface Mount |
Height (English) | 10 in. |
Height (Metric) | 25.4 cm |
Description
- Bright colors and bold text provide high visibility to critical messages
- Sign reads “DANGER NITROGEN”
- Black/red on white
- Designed to withstand harsh industrial environments
- Available in different sizes and materials
B-401
- Plastic — high-impact polystyrene
- Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic
Specifications
ANSI Z535, OSHA 1910.145 | |
Surface Mount | |
25.4 cm | |
14 in. | |
DANGER Nitrogen | |
Red on White |
1 Ea. | |
10 in. | |
-40°F | |
35.56 cm | |
Chemical and Hazardous Materials Sign |