missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Danger: Combustible Liquids Signs
Danger: Combustible Liquids Signs are extremely durable B-120 Premium Fiberglass that will not chip, fade, rust, shatter or peel for up to 25 years.
$15.19 - $15.19
Specifications
Quantity | 1 Ea. |
---|---|
Mounting Type | Surface Mount |
Min. Temperature (English) | -40°F |
Displayed Text | DANGER Combustible Liquids |
Product Type | Chemical and Hazardous Materials Sign |
Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Price | Quantity | |||
19-102-398
|
Brady™
22317 |
Plastic | 7 in. | 10 in. |
Each for $15.19
|
|
|||
Description
- Graphics are permanently embedded within the sign panel, allowing for use in the most severe environments
- Excellent chemical and abrasion resistance
- Ideal for both indoor and outdoor applications
- Service temperature of -40°F to 193°F (-40°C to 90°C)
Specifications
1 Ea. | |
-40°F | |
Chemical and Hazardous Materials Sign |
Surface Mount | |
DANGER Combustible Liquids |