missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Danger: Caustic Signs
Identify equipment, rooms and materials that contain or are contaminated with hazardous agents.
$14.58 - $37.51
Specifications
Quantity | 1 Ea. |
---|---|
Mounting Type | Surface Mount |
Min. Temperature (English) | -40°F |
Displayed Text | DANGER Caustic |
Product Type | Chemical and Hazardous Materials Sign |
Catalog Number | Mfr. No. | Material | Height (English) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Height (English) | Price | Quantity | ||||
19-016886
|
Brady™
BRD72392 |
Fiberglass | 7 in. |
Each for $26.44
|
|
||||
19-035-466
|
Brady™
84346 |
Polyester | 10 in. |
Each for $37.51
|
|
||||
19-102-103
|
Brady™
22306 |
Plastic | 10 in. |
Each for $21.84
|
|
||||
19-102-104
|
Brady™
84345 |
Polyester | 7 in. |
Each for $14.58
|
|
||||
Description
- Bright colors and bold text provide high visibility to critical messages
- Black/red on white
- Sign reads “DANGER CAUSTIC”
- Designed to withstand harsh industrial environments
- Available in different sizes and materials
B-401
- Plastic — high-impact polystyrene
- Premium fiberglass — fiberglass reinforced polyester
- Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic
Specifications
1 Ea. | |
-40°F | |
Chemical and Hazardous Materials Sign |
Surface Mount | |
DANGER Caustic | |
Red on White |