missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, 4% (w/v) with Indicator (Bromocresol Green-Methyl Red), Certified, 4.0% (w/v) ±0.2%, LabChem™
$79.00 - $290.20
Chemical Identifiers
CAS | 10043-35-3 |
---|---|
Molecular Formula | BH3O3 |
Molecular Weight (g/mol) | 61.83 |
MDL Number | MFCD00011337 |
InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
Synonym | orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe |
PubChem CID | 7628 |
ChEBI | CHEBI:33118 |
IUPAC Name | boric acid |
SMILES | OB(O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
LC117502
|
LabChem
LC117502 |
1 L | Poly Bottle |
Each for $79.00
|
|
||||
LC117504
|
LabChem
LC117504 |
4 L | Poly Bottle |
Each for $136.60
|
|
||||
LC117506
|
LabChem
LC117506 |
10 L | Cubitainer™ |
Each for $218.33
|
|
||||
LC117505
|
LabChem
LC117505 |
20 L | Cubitainer™ |
Each for $290.20
|
|
||||
Chemical Identifiers
10043-35-3 | |
61.83 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
7628 | |
boric acid |
BH3O3 | |
MFCD00011337 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe | |
CHEBI:33118 | |
OB(O)O |
Specifications
10043-35-3,7732-18-5,62625-32-5,845-10-3 | |
BH3O3 | |
MFCD00011337 | |
Soluble in water | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
boric acid | |
7628 | |
61.83 | |
Certified | |
Boron | |
1 L | |
Boric Acid, 4% (w/v) with Indicator (Bromocresol Green-Methyl Red) |
4,95.998,0.001,0.001 | |
H3BO3 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, Tris-borate-EDTA, h3bo3, flea prufe | |
Passes Test | |
OB(O)O | |
61.83 | |
CHEBI:33118 | |
4.0% ±0.2% (w/v) | |
Poly Bottle | |
Red | |
Liquid |
Safety and Handling
GHS H Statement
May damage fertility or the unborn child.
GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature