Learn More
Tasco™ Blackhawk Ear Muff
Blackhawk 3 position headband, dielectric, NRR 27, CSA Class A
Supplier: Tasco™ DTC772700
Description
- Dielectric, three position ear muff
- 27 NRR over-the-head, 26 NRR under-chin/behind-neck
- Tested at an independent NVLAP accredited laboratory
- Soft-Seal™ ear cushions provide exceptional comfort and fit
- Special foam head pad for all day comfort
- Sculpted headband with molded grips for easy cup adjustment
- Consistent full spectrum attenuation
Specifications
Black | |
Class A | |
2700 | |
Black | |
Multi-position |
Yes | |
Black | |
27 NRR (Over-the-head) 26 NRR (Under-chin/Behind-neck) | |
Yes | |
No |