missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(triphenylphosphine)nickel(II) Dichloride 96.0+%, TCI America™
Supplier: TCI America B1571100G
Specifications
Bis(triphenylphosphine)nickel(II) Dichloride | |
Black-Green | |
MFCD00009592 MFCD00003043 MFCD20489348 | |
bis triphenylphosphine dichloronickel ii | |
Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
654.18 | |
654.18 | |
Crystalline Powder |
14264-16-5 | |
C36H30Cl2NiP2 | |
100 g | |
VHQPRJDILRYZBG-UHFFFAOYSA-L | |
dichloronickelbis(ylium); bis(triphenylphosphane) | |
131664342 | |
≥96.0% (T) |
Chemical Identifiers
14264-16-5 | |
654.18 | |
VHQPRJDILRYZBG-UHFFFAOYSA-L | |
131664342 | |
Cl[Ni++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C36H30Cl2NiP2 | |
MFCD00009592 MFCD00003043 MFCD20489348 | |
bis triphenylphosphine dichloronickel ii | |
dichloronickelbis(ylium); bis(triphenylphosphane) |
Safety and Handling
RTECSNumber : QR6170000
TSCA : Yes