Learn More
Bis(tetramethylcyclopentadienyl)nickel(II), 98+%
CAS: 79019-60-6 | C18H26Ni | 301.099 g/mol
$564.90 - $564.90
Chemical Identifiers
CAS | 79019-60-6 |
---|---|
Molecular Formula | C18H26Ni |
Molecular Weight (g/mol) | 301.099 |
MDL Number | MFCD01862460 |
InChI Key | PPBIVTWBQQUEKM-UHFFFAOYSA-N |
Synonym | nickel 2+ bis 2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide, nickel 2+ ion bis 2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide |
PubChem CID | 73994274 |
IUPAC Name | nickel(2+);1,2,3,5-tetramethylcyclopenta-1,3-diene |
SMILES | CC1[C-]=C(C(=C1C)C)C.CC1[C-]=C(C(=C1C)C)C.[Ni+2] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2778403
|
Thermo Scientific Chemicals
H2778403 |
1 g |
Each for $564.90
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79019-60-6 | |
301.099 | |
PPBIVTWBQQUEKM-UHFFFAOYSA-N | |
73994274 | |
CC1[C-]=C(C(=C1C)C)C.CC1[C-]=C(C(=C1C)C)C.[Ni+2] |
C18H26Ni | |
MFCD01862460 | |
nickel 2+ bis 2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide, nickel 2+ ion bis 2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide | |
nickel(2+);1,2,3,5-tetramethylcyclopenta-1,3-diene |
Specifications
79019-60-6 | |
C18H26Ni | |
1 g | |
nickel 2+ bis 2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide, nickel 2+ ion bis 2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide | |
CC1[C-]=C(C(=C1C)C)C.CC1[C-]=C(C(=C1C)C)C.[Ni+2] | |
301.099 | |
301.1 | |
Bis(tetramethylcyclopentadienyl)nickel(II) |
107°C to 112°C | |
MFCD01862460 | |
Air and moisture sensitive | |
PPBIVTWBQQUEKM-UHFFFAOYSA-N | |
nickel(2+);1,2,3,5-tetramethylcyclopenta-1,3-diene | |
73994274 | |
≥98% |
Safety and Handling
GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only