missing translation for 'onlineSavingsMsg'
Learn More

BIN1 Monoclonal Antibody (K70035_11G6), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Fisher Chemical MA555637

 View more versions of this product

Catalog No. MA555637


Only null left
Explore available promotions

Description

Description

Sequence of this protein is as follows: AGKIASNVQK KLTRAQEKVL QKLGKADETK DEQFEQCVQN FNKQLTEGTR LQKDLRTYLA SVKAMHEASK KLNECLQEVY EPDWPGRDEA NKIAENNDLL WMDYHQKLVD QALLTMDTYL GQFPDIKSRI AKRGRKLVDY DSARHHYESL QTAKKKDEAK IAKPVSLLEK AAPQWCQGKL QAHLVAQTNL LRNQAEEELI KAQKVFEEMN VDLQEELPSL WNSRVGFYVN TFQSIAGLEE NFHKEMSKLN QNLNDVLVGL EKQHGSNTFT

BIN1 Monoclonal antibody specifically detects BIN1 in Human, Rat samples. It is validated for Immunoprecipitation, Western Blot
Specifications

Specifications

BIN1
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
O00499, O08839
Mouse
Protein A
RUO
117028, 274
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunoprecipitation, Western Blot
K70035_11G6
Unconjugated
BIN1
ALP-1; AMPH 2; Amph2; amphiphysin 2; amphiphysin II; amphiphysin IIamph2; amphiphysin-like; amphiphysin-like protein; AMPHL; Bin1; box dependant MYC interacting protein 1; box-dependent myc-interacting protein 1; BRAMP-2; bridging integrator 1; DKFZp547F068; MGC10367; MGC105358; myc box dependent interacting protein 1; myc box-dependent-interacting protein 1; SH3 domain-containing protein 9; SH3P9; similar to bridging integrator 1 isoform 9
Recombinant human BIN1 (immunogen range is 11-280).
50 μg
Primary
Human, Rat
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions