missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betulinic Acid 97.0+%, TCI America™
$259.11 - $1193.00
Chemical Identifiers
CAS | 472-15-1 |
---|---|
Molecular Formula | C30H48O3 |
Molecular Weight (g/mol) | 456.711 |
InChI Key | QGJZLNKBHJESQX-FZFNOLFKSA-N |
Synonym | betulinic acid, mairin, betulic acid, ccris 6748, 3-hydroxylup-20 29-en-28-oic acid, 3beta-hydroxy-20 29-lupaene-28-oic acid, 3beta-hydroxy-lup-20 29-en-28-oic acid, lup-20 29-en-28-oic acid, 3-hydroxy-, 3beta, 1r,3as,5ar,5br,7ar,9s,11ar,11br,13ar,13br-9-hydroxy-1-isopropenyl-5a,5b,8,8,11a-pentamethyl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta a chrysene-3a-carboxylic acid, 1r,3as,5ar,5br,7ar,9s,11ar,11br,13ar,13br-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta a chrysene-3a-carboxylic acid |
PubChem CID | 64971 |
ChEBI | CHEBI:3087 |
IUPAC Name | (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta[a]chrysene-3a-carboxylic acid |
SMILES | CC(=C)C1CCC2(C1C3CCC4C5(CCC(C(C5CCC4(C3(CC2)C)C)(C)C)O)C)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B2836100MG
|
TCI America
B2836100MG |
100 mg |
Each for $259.11
|
|
|||||
B28361G
|
TCI America
B28361G |
1 g |
Each for $1,193.00
|
|
|||||
Chemical Identifiers
472-15-1 | |
456.711 | |
betulinic acid, mairin, betulic acid, ccris 6748, 3-hydroxylup-20 29-en-28-oic acid, 3beta-hydroxy-20 29-lupaene-28-oic acid, 3beta-hydroxy-lup-20 29-en-28-oic acid, lup-20 29-en-28-oic acid, 3-hydroxy-, 3beta, 1r,3as,5ar,5br,7ar,9s,11ar,11br,13ar,13br-9-hydroxy-1-isopropenyl-5a,5b,8,8,11a-pentamethyl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta a chrysene-3a-carboxylic acid, 1r,3as,5ar,5br,7ar,9s,11ar,11br,13ar,13br-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta a chrysene-3a-carboxylic acid | |
CHEBI:3087 | |
CC(=C)C1CCC2(C1C3CCC4C5(CCC(C(C5CCC4(C3(CC2)C)C)(C)C)O)C)C(=O)O |
C30H48O3 | |
QGJZLNKBHJESQX-FZFNOLFKSA-N | |
64971 | |
(1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta[a]chrysene-3a-carboxylic acid |
Specifications
472-15-1 | |
C30H48O3 | |
betulinic acid, mairin, betulic acid, ccris 6748, 3-hydroxylup-20 29-en-28-oic acid, 3beta-hydroxy-20 29-lupaene-28-oic acid, 3beta-hydroxy-lup-20 29-en-28-oic acid, lup-20 29-en-28-oic acid, 3-hydroxy-, 3beta, 1r,3as,5ar,5br,7ar,9s,11ar,11br,13ar,13br-9-hydroxy-1-isopropenyl-5a,5b,8,8,11a-pentamethyl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta a chrysene-3a-carboxylic acid, 1r,3as,5ar,5br,7ar,9s,11ar,11br,13ar,13br-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta a chrysene-3a-carboxylic acid | |
CC(=C)C1CCC2(C1C3CCC4C5(CCC(C(C5CCC4(C3(CC2)C)C)(C)C)O)C)C(=O)O | |
456.711 | |
CHEBI:3087 | |
≥97.0% (GC,T) | |
Betulinic Acid |
294°C | |
100 mg | |
QGJZLNKBHJESQX-FZFNOLFKSA-N | |
(1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-hydroxy-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta[a]chrysene-3a-carboxylic acid | |
64971 | |
456.71 | |
Crystalline Powder |
Safety and Handling
TSCA : No