missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Beta Cyanoethyl Fastphoramidite™, dG(dmf)
The dG(dmf) Beta Cyanoethyl Fastphoramidite™ is used as a component of a fast deprotecting chemistry set including dA(Bz), dC(Bz) and T. FastPhoramidite™ series deprotects in 1 hour at 65°C.
Supplier: Applied Biosystems™ 401165
Description
High QualityOur phosphoramidites benefit from proprietary manufacturing processes that yield high-purity products. In synthesis they perform at a correspondingly high level, producing pure, High Quality DNA at all scales. We guarantee that 5'-dimethoxytrityl nucleoside β-cyanoethyl phosphoramidites, monomer intermediates for licensed β-cyanoethyl phosphoramidite chemistry, will produce DNA that is at least 99.8% chemically authentic when used with recommended synthesis cycles.Fast ResultsFastPhoramidite™ reagents offer fast deprotection for primer-length oligonucleotides. Simply substituting Fast G, dG(dmf), for our standard dG(iBu) shortens deprotection at 55°C from eight hours to two. Elevating the temperature to 65°C further cuts deprotection to 1 hour.Dry phosphoroamidites (powder form) are guaranteed for one year from the date of shipment. Lifetime of diluted phosphoramidites in residence on the instrument is two weeks when used with CPG synthesis supports, and three weeks when used with polystyrene supports.β-cyanoethyl phosphoramidites are for use on ABI synthesizers to guarantee you high quality nucleic acid synthesis.
- Proprietary manufacturing processes yield high-purity β-cyanoethyl phosphoramidites, which in turn guarantee production of high-quality DNA at all scales.
- FastPhoramidite™ reagents deprotect in one hour at 65°C to shorten overall oligonucleotide production times and costs.
- β-cyanoethyl phosphoramidites are use-tested and optimized for use on the ABI synthesizers to guarantee you high-quality nucleic acid synthesis.
Specifications
Powder | |
dG(dmf) beta CEP | |
Deoxyguanosine (dG) | |
Room Temperature | |
Phosphoramidite |
3900 DNA Synthesizer | |
Store in freezer (-5 to -30°C). | |
Fast deprotection | |
2 g | |
DNA |
For Research Use Only. Not for use in diagnostic procedures.