missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHOWA™ 7714 Series PVC Coated General Purpose Gloves, Cotton Jersey Liner
General purpose gloves with 14 in. extended forearm length protects against a range of hazards including chemicals, oil and grease.
$8.44 - $8.44
Specifications
Certifications/Compliance | CFIA |
---|---|
Color | Black |
Latex Free | Yes |
Material | PVC |
Size | Large |
Catalog Number | Mfr. No. | Grip Design | Series | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Grip Design | Series | Price | Quantity | ||||
11-395-24
|
SHOWA™
7714R10 |
Rough | 7714R Series |
Pair for $8.44
Case of 72 Pair for $546.48
|
|
||||
Description
Both styles are ideal for working with chemicals, oil, or grease in refining, mining, automotive subassembly, or construction
- Fully coated PVC with smooth or rough finish
- Cotton jersey lined Sanitized™
- Recyclable
- Washable
Specifications
CFIA | |
Yes | |
Large | |
0.55 mm | |
Gauntlet | |
Lined | |
35.5 cm |
Black | |
PVC | |
22 mil | |
Chemical spray and treatment, coating preparation, laboratory, pharma and analysis, painting and spray, washing and cleaning | |
Cotton | |
14 in. |