missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bergenin 98.0+%, TCI America™
$322.48 - $706.48
Chemical Identifiers
CAS | 477-90-7 |
---|---|
Molecular Formula | C14H16O9 |
Molecular Weight (g/mol) | 328.273 |
MDL Number | MFCD00133120 |
InChI Key | YWJXCIXBAKGUKZ-HJJNZUOJSA-N |
PubChem CID | 66065 |
ChEBI | CHEBI:69499 |
IUPAC Name | (2R,3S,4S,4aR,10bS)-3,4,8,10-tetrahydroxy-2-(hydroxymethyl)-9-methoxy-3,4,4a,10b-tetrahydro-2H-pyrano[3,2-c]isochromen-6-one |
SMILES | COC1=C(C=C2C(=C1O)C3C(C(C(C(O3)CO)O)O)OC2=O)O |
Chemical Identifiers
477-90-7 | |
328.273 | |
YWJXCIXBAKGUKZ-HJJNZUOJSA-N | |
CHEBI:69499 | |
COC1=C(C=C2C(=C1O)C3C(C(C(C(O3)CO)O)O)OC2=O)O |
C14H16O9 | |
MFCD00133120 | |
66065 | |
(2R,3S,4S,4aR,10bS)-3,4,8,10-tetrahydroxy-2-(hydroxymethyl)-9-methoxy-3,4,4a,10b-tetrahydro-2H-pyrano[3,2-c]isochromen-6-one |
Specifications
240°C | |
White | |
MFCD00133120 | |
COC1=C(C=C2C(=C1O)C3C(C(C(C(O3)CO)O)O)OC2=O)O | |
328.273 | |
CHEBI:69499 | |
≥98.0% (HPLC) | |
Crystalline Powder |
477-90-7 | |
C14H16O9 | |
YWJXCIXBAKGUKZ-HJJNZUOJSA-N | |
(2R,3S,4S,4aR,10bS)-3,4,8,10-tetrahydroxy-2-(hydroxymethyl)-9-methoxy-3,4,4a,10b-tetrahydro-2H-pyrano[3,2-c]isochromen-6-one | |
66065 | |
328.27 | |
1 g | |
Bergenin |
Safety and Handling
RTECSNumber : UQ0337200
TSCA : No
Recommended Storage : Refrigerator