missing translation for 'onlineSavingsMsg'
Learn More

Bergamot Oil, Natural, Spectrum™ Chemical

BE161, 8007-75-8

$246.09 - $11778.40

Chemical Identifiers

CAS 8007-75-8
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18600926
View Documents
Spectrum Chemical Mfg Cor
BE16125ML
25 mL Amber Glass Bottle
Each for $246.09
Only null left
 
18600927
View Documents
Spectrum Chemical Mfg Cor
BE161100ML
100 mL Amber Glass Bottle
Each for $604.34
Only null left
 
18600928
View Documents
Spectrum Chemical Mfg Cor
BE161500MLGL
500 mL Amber Glass Bottle
Each for $2,022.17
Only null left
 
18600929
View Documents
Spectrum Chemical Mfg Cor
BE1615KG
5 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Bergamot Oil, Natural , also known as Bergamot essential oil, is used as a top note in several perfumes. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications

Chemical Identifiers

8007-75-8

Specifications

8007-75-8
Amber Glass Bottle
UN1169
100%
25 mL
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'