missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, ACS, 99%, Spectrum™ Chemical

$295.83 - $29808.73

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18609053
View Documents
Spectrum Chemical Mfg Cor
B1546100ML
100 mL Steel Can
Each for $295.83
Only null left
 
18609052
View Documents
Spectrum Chemical Mfg Cor
B1546500MLGL
500 mL Steel Can
Each for $703.37
Only null left
 
18609056
View Documents
Spectrum Chemical Mfg Cor
B15464LT
4 L Steel Can
Each for $2,418.50
Only null left
 
18609050
View Documents
Spectrum Chemical Mfg Cor
B154620LT
20 L Steel Can
Each for $6,801.67
Only null left
 
18609051
View Documents
Spectrum Chemical Mfg Cor
B1546200LTBL
200 L Steel Can N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

100-51-6
108.14
phenylmethanol
C7H8O
WVDDGKGOMKODPV-UHFFFAOYSA-N
OCC1=CC=CC=C1

Specifications

100-51-6
205.3°C
Steel Can
100 mL
OCC1=CC=CC=C1
108.14
ACS
1
5e-05
C7H8O
WVDDGKGOMKODPV-UHFFFAOYSA-N
phenylmethanol
0.99
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.