missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoyl Bromide 98.0+%, TCI America™
$274.02 - $274.02
Chemical Identifiers
CAS | 618-32-6 |
---|---|
Molecular Formula | C7H5BrO |
Molecular Weight (g/mol) | 185.02 |
MDL Number | MFCD00000112 |
InChI Key | AQIHMSVIAGNIDM-UHFFFAOYSA-N |
Synonym | benzoylbromid, benzoylbromide, bzbr, benzoic acid bromide, acmc-209mwn, benzoyl bromide, benzbromarone impurity a, 6ci,7ci,8ci,9ci, benzoic acid bromide;benzenecarbonyl bromide |
PubChem CID | 12056 |
IUPAC Name | benzoyl bromide |
SMILES | BrC(=O)C1=CC=CC=C1 |
Chemical Identifiers
618-32-6 | |
185.02 | |
AQIHMSVIAGNIDM-UHFFFAOYSA-N | |
12056 | |
BrC(=O)C1=CC=CC=C1 |
C7H5BrO | |
MFCD00000112 | |
benzoylbromid, benzoylbromide, bzbr, benzoic acid bromide, acmc-209mwn, benzoyl bromide, benzbromarone impurity a, 6ci,7ci,8ci,9ci, benzoic acid bromide;benzenecarbonyl bromide | |
benzoyl bromide |
Specifications
618-32-6 | |
219°C | |
MFCD00000112 | |
3265 | |
AQIHMSVIAGNIDM-UHFFFAOYSA-N | |
benzoyl bromide | |
12056 | |
≥98.0% (GC,T) | |
Benzoyl Bromide |
Yellow | |
C7H5BrO | |
25 g | |
benzoylbromid, benzoylbromide, bzbr, benzoic acid bromide, acmc-209mwn, benzoyl bromide, benzbromarone impurity a, 6ci,7ci,8ci,9ci, benzoic acid bromide;benzenecarbonyl bromide | |
BrC(=O)C1=CC=CC=C1 | |
185.02 | |
185.02 | |
Liquid |
Safety and Handling
TSCA : Yes