Learn More
Benzopinacol, 98%
CAS: 464-72-2 | C26H22O2 | 366.46 g/mol
$119.52 - $119.52
Chemical Identifiers
CAS | 464-72-2 |
---|---|
Molecular Formula | C26H22O2 |
Molecular Weight (g/mol) | 366.46 |
MDL Number | MFCD00004448 |
InChI Key | MFEWNFVBWPABCX-UHFFFAOYSA-N |
Synonym | benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl |
PubChem CID | 94766 |
IUPAC Name | 1,1,2,2-tetraphenylethane-1,2-diol |
SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1142114
|
Thermo Scientific Chemicals
A1142114 |
25 g |
Each for $119.52
|
|
|||||
Description
Benzopinacol is a catalyst of the formation of unsaturated polyesters. It is also used as an initiator of polymerisation by free radicals. It is also used as an organic synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications: Benzopinacol is a catalyst of the formation of unsaturated polyesters. It is also used as an initiator of polymerisation by free radicals. It is also used as an organic synthesis intermediate.Chemical Identifiers
464-72-2 | |
366.46 | |
MFEWNFVBWPABCX-UHFFFAOYSA-N | |
94766 | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O |
C26H22O2 | |
MFCD00004448 | |
benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl | |
1,1,2,2-tetraphenylethane-1,2-diol |
Specifications
464-72-2 | |
C26H22O2 | |
25 g | |
14,1100 | |
It is soluble in ethanol,ether,acetone insoluble in carbon disulfide and water. | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O | |
366.46 | |
366.46 | |
Benzopinacol |
174°C to 180°C | |
MFCD00004448 | |
2061947 | |
benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl | |
MFEWNFVBWPABCX-UHFFFAOYSA-N | |
1,1,2,2-tetraphenylethane-1,2-diol | |
94766 | |
98% |
Safety and Handling
EINECSNumber : 207-356-8
RTECSNumber : KI2750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only