missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid, Crystal, EP, BP, JP, USP, 99.5-100.5%, Spectrum™ Chemical

BE140, 65-85-0, C7H6O2

$545.23 - $8215.51

Chemical Identifiers

CAS 65-85-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
InChI Key WPYMKLBDIGXBTP-UHFFFAOYSA-N
IUPAC Name benzoic acid
SMILES OC(=O)C1=CC=CC=C1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18600902
View Documents
Spectrum Chemical Mfg Cor
BE140500GM
500 g Poly Pail
Each for $545.23
Only null left
 
18600903
View Documents
Spectrum Chemical Mfg Cor
BE14025KG
2.5 kg Poly Pail
Each for $1,486.34
Only null left
 
18600904
View Documents
Spectrum Chemical Mfg Cor
BE14012KG
12 kg Poly Pail
Each for $3,966.46
Only null left
 
18600906
View Documents
Spectrum Chemical Mfg Cor
BE14050KGBL
50 kg Poly Pail
Each for $8,215.51
Only null left
 
Description

Description

Spectrum™ Chemical Benzoic Acid, Crystal, USP, EP, BP, JP has antifungal and topical antiseptic properties. Spectrum Chemical USP, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

65-85-0
122.12
benzoic acid
C7H6O2
WPYMKLBDIGXBTP-UHFFFAOYSA-N
OC(=O)C1=CC=CC=C1

Specifications

65-85-0
0.05%
Poly Pail
500 g
OC(=O)C1=CC=CC=C1
122.12
Multi Compendia
100%
0.5%
C7H6O2
WPYMKLBDIGXBTP-UHFFFAOYSA-N
benzoic acid
99.5 to 100.5%
Crystalline Powder or Lumps
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'