missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bentonite, acid-leached, Thermo Scientific™
$35.46 - $331.44
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC471231000
|
Thermo Scientific Chemicals
471231000 |
100 g |
Each for $35.46
|
|
|||||
AC471230010
|
Thermo Scientific Chemicals
471230010 |
1 kg |
Each for $103.92
|
|
|||||
AC471230050
|
Thermo Scientific Chemicals
471230050 |
5 kg |
Each for $331.44
|
|
|||||
Chemical Identifiers
70131-50-9 |
Specifications
0.03 mg/KOH/g (Free Acidity) | |
Conforms to structure | |
100 g | |
Granules |
70131-50-9 | |
Plastic Bottle | |
≤ 0.1 mg/KOH/g (Free Acidity) | |
Bentonite |
Safety and Handling
ShelfLife : 5 years
EINECSNumber : 274-324-8
Recommended Storage : Normal conditions