missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Benidipine hydrochloride

Chemical Identifiers

CAS 91599-74-5
Molecular Formula C28H32ClN3O6
Molecular Weight (g/mol) 542.03
InChI Key KILKDKRQBYMKQX-MIPPOABVSA-N
IUPAC Name hydrogen 3-(3R)-1-benzylpiperidin-3-yl 5-methyl (4R)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate chloride
SMILES [H+].[Cl-].COC(=O)C1=C(C)NC(C)=C([C@@H]1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)O[C@@H]1CCCN(CC2=CC=CC=C2)C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC465100010
View Documents
Thermo Scientific™
465100010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC465102500
View Documents
Thermo Scientific™
465102500
250 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

91599-74-5
542.03
hydrogen 3-(3R)-1-benzylpiperidin-3-yl 5-methyl (4R)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate chloride
C28H32ClN3O6
KILKDKRQBYMKQX-MIPPOABVSA-N
[H+].[Cl-].COC(=O)C1=C(C)NC(C)=C([C@@H]1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)O[C@@H]1CCCN(CC2=CC=CC=C2)C1

Specifications

91599-74-5
1 g
[H+].[Cl-].COC(=O)C1=C(C)NC(C)=C([C@@H]1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)O[C@@H]1CCCN(CC2=CC=CC=C2)C1
542.03
Benidipine hydrochloride
C28H32ClN3O6
KILKDKRQBYMKQX-MIPPOABVSA-N
hydrogen 3-(3R)-1-benzylpiperidin-3-yl 5-methyl (4R)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate chloride
542.02
Videos
Safety and Handling

Safety and Handling

GHS H Statement Toxic if swallowed.

Suspected of damaging fertility or the unborn child.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only