missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bendazac, Thermo Scientific™
Chemical Identifiers
CAS | 20187-55-7 |
---|---|
Molecular Formula | C16H13N2NaO3 |
Molecular Weight (g/mol) | 304.28 |
InChI Key | GHFGHIKJGVMEFT-UHFFFAOYSA-M |
IUPAC Name | sodium 2-[(1-benzyl-1H-indazol-3-yl)oxy]acetate |
SMILES | [Na+].[O-]C(=O)COC1=NN(CC2=CC=CC=C2)C2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC464920010
|
Thermo Scientific Chemicals
464920010 |
1 g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC464920050
|
Thermo Scientific Chemicals
464920050 |
5 g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
20187-55-7 | |
304.28 | |
sodium 2-[(1-benzyl-1H-indazol-3-yl)oxy]acetate |
C16H13N2NaO3 | |
GHFGHIKJGVMEFT-UHFFFAOYSA-M | |
[Na+].[O-]C(=O)COC1=NN(CC2=CC=CC=C2)C2=CC=CC=C12 |
Specifications
20187-55-7 | |
1 g | |
GHFGHIKJGVMEFT-UHFFFAOYSA-M | |
sodium 2-[(1-benzyl-1H-indazol-3-yl)oxy]acetate | |
282.3 |
C16H13N2NaO3 | |
Solubility aq. soln.: insoluble. Other solubilities: soluble in chloroform, ethanol, acetone | |
[Na+].[O-]C(=O)COC1=NN(CC2=CC=CC=C2)C2=CC=CC=C12 | |
304.28 | |
Bendazac |
Safety and Handling
GHS H Statement Harmful if swallowed.
GHS Signal Word: Warning
EINECSNumber : 243-569-2
RUO â Research Use Only