Learn More
Bel-Art™ Cover Glass Forceps
Tips shaped to easily lift cover glass. Bel-Art™ Cover Glass Forceps are fully coated with brown Teflon™ FEP and comply with FDA regulations concerning direct food contact.
$253.52 - $254.38
Catalog Number | Mfr. No. | Length (Metric) | Tip Style | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Tip Style | Price | Quantity | ||||
22-124370
|
Bel-Art™
H379430000 |
114 mm | Straight |
Pack of 2 Each for $254.38
Case of 24 Pack for $6,105.12
|
|
||||
22-124-586
|
Bel-Art™
H379420000 |
102 mm | Bent |
Pack of 2 Each for $253.52
Case of 24 Pack for $6,084.48
|
|
||||
Description
- Brown PTFE FEP coating guards against scratches
- Wide, bent tip style is perfect for lifting cover glasses
Specifications
Forceps |