missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: m LES Endo Agar

For enumerating coliforms in water by membrane filtration

$330.66 - $330.66

Specifications

Certifications/Compliance CE, COMPF, SMD, SMWW
Content And Storage <30°C
Form Dehydrated
Media Type mLES Endo Agar
Product Type Agar
View More Specs

Products 1
Catalog Number Mfr. No. Quantity For Use With (Application) Price Quantity  
Catalog Number Mfr. No. Quantity For Use With (Application) Price Quantity  
DF0736-17-2
View Documents
BD
273620
500 g Coliform enumeration
Each for $330.66
Only null left
 
Description

Description

  • Formulation: Yeast extract 1.2g/L, Casitone 3.7g/L, Thiopeptone 3.7g/L, Tryptose 7.5g/L, Lactose 9.4g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 3.7g/L, Sodium Desoxycholate 0.1g/L, Sodium Lauryl Sulfate 0.05g/L, Sodium Sulfite 1.6g

Certifications

CE, COMPF, SMD, SMWW

Specifications

Specifications

CE, COMPF, SMD, SMWW
Dehydrated
Agar
Coliform enumeration
<30°C
mLES Endo Agar
Yeast extract 1.2g/L, Casitone 3.7g/L, Thiopeptone 3.7g/L, Tryptose 7.5g/L, Lactose 9.4g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 3.7g/L, Sodium Desoxycholate 0.1g/L, Sodium Lauryl Sulfate 0.05g/L, Sodium Sulfite 1.6g/L, Basic Fuchsin 0.8g/L, Agar 15g/L
Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'