missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: Trypticase™ Soy Agar (Soybean-Casein Digest Agar)
Used for isolation and cultivation of non-fastidious and fastidious microorganisms
Supplier: BD 221283
Description
- Ready for immediate use
- Not used for anaerobes
Specifications
ISO/DIS, 11014 | |
2°C to 8°C | |
Trypticase Soy Agar | |
Cultivation of Non-fastidious and fastidious microorganisms (not for anaerobes) | |
100/Pk. |
Light Beige | |
United States | |
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L | |
Plate | |
100 x 15 mm |