missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: Standard Methods Agar (Plate Count Agar)
For the isolation of microorganisms from samples or specimens
Supplier: BD 297030
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
ISO/DIS, 11014 | |
Plate | |
Standard methods Agar (Plate count agar) |
Beige | |
United States | |
20/Pk. |