missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Prepared Plated Media: Mueller Hinton II Agar
For antimicrobial disc diffusion susceptibility testing of common, rapidly growing bacteria by the Bauer-Kirby method
$164.61 - $240.23
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Color | Beige |
Content And Storage | 2°C to 8°C |
Form | Solid |
Manufacturing Origin | United States |
Description
- Ready-to-use
- 100 x 10mm, 100 x 15mm and 150 x 15mm-style plates
Specifications
ISO/DIS, 11014 | |
2°C to 8°C | |
United States | |
Beef extract 2g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L, Agar 17g/L |
Beige | |
Solid | |
Mueller Hinton II Agar | |
Antimicrobial disc diffusion susceptibility testing of common rapidly growing bacteria by the Bauer-Kirby method |