missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bathophenanthroline, For Spectrophotometric Det. of Fe in Serum, ≥99.0%, MilliporeSigma™ Supelco™
Bathophenanthroline (BPhen) is a chelating agent and is specific for ferrous ions.
$265.96 - $1026.71
Chemical Identifiers
MDL Number | MFCD00004976 |
---|---|
Synonym | 4,7-Diphenyl-1,10-phenanthroline; BPhen |
Chemical Identifiers
MFCD00004976 | |
4,7-Diphenyl-1,10-phenanthroline; BPhen |
Specifications
218°C to 220°C; 218°C to 222°C | |
C24H16N2 | |
1 g | |
332.40 |
1662-01-7 | |
MFCD00004976 | |
4,7-Diphenyl-1,10-phenanthroline; BPhen | |
≥99.0% (NT); ≥99.0% |
Safety and Handling
RTECSNumber : SF8427000