missing translation for 'onlineSavingsMsg'
Learn More

Barium hydroxide octahydrate, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥98%, Honeywell Fluka™

Puriss. p.a., Reag. ISO, Reag. Ph. Eur.,≥98%

$175.10 - $794.10

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
PubChem CID 17749109
IUPAC Name barium(2+);dihydroxide;octahydrate
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6000367
View Documents
Honeywell Chemicals
31127100G
100 g
Each for $175.10
Only null left
 
6000369
View Documents
Honeywell Chemicals
31127500G
500 g
Each for $403.05
Only null left
 
6000368
View Documents
Honeywell-Fluka
311271KG
1 kg
Each for $794.10
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

12230-71-6
315.46
ZUDYPQRUOYEARG-UHFFFAOYSA-L
17749109
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
BaH18O10
MFCD00149152
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
barium(2+);dihydroxide;octahydrate

Specifications

12230-71-6
BaH18O10
MFCD00149152
barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
315.46
315.46g/mol
puriss. p.a.
Barium Hydroxide Octahydrate, Reag. Ph. Eur. reag. ISO
100 g
Ba(OH)2 · 8H2O
3262
ZUDYPQRUOYEARG-UHFFFAOYSA-L
barium(2+);dihydroxide;octahydrate
17749109
≥98%
Plastic Bottle
Videos
Safety and Handling

Safety and Handling

P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

H302 + H332-H314

EINECSNumber : 241-234-5

SDS
missing translation for 'documents'

missing translation for 'documents'