Learn More
Perkin Elmer Canada BTO Orange Injection Port Septa, Pkg. 50
Supplier: Perkin Elmer Canada N9302972
Septa rated to 400 C. The precision molded silicone rubber septa BTO (Bleed Temperature Optimized) are premium ultra-low bleed injector septa for todays most demanding applications.