missing translation for 'onlineSavingsMsg'
Learn More

Azoic Diazo Component 24 (Salt) 95.0+%, TCI America™

$85.75 - $221.44

Chemical Identifiers

CAS 55663-99-5
Molecular Formula C30H28Cl4N6O6Zn
Molecular Weight (g/mol) 775.77
MDL Number MFCD00074762
InChI Key LLARVKAOQLUCQI-UHFFFAOYSA-L
Synonym 4-Benzamido-2,5-dimethoxybenzenediazonium Salt Zinc Chloride, Fast Blue RR Salt
PubChem CID 134159107
IUPAC Name zinc;N-(4-diazonio-2,5-dimethoxyphenyl)benzenecarboximidate;dichloride;dihydrochloride
SMILES COC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OC)[N+]#N.COC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OC)[N+]#N.Cl.Cl.[Cl-].[Cl-].[Zn+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B07855G
View Documents
TCI America
B07855G
5 g
Each for $85.75
Only null left
 
B078525G
View Documents
TCI America
B078525G
25 g
Each for $221.44
Only null left
 
Description

Description

Specifications

Chemical Identifiers

55663-99-5
775.77
LLARVKAOQLUCQI-UHFFFAOYSA-L
134159107
COC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OC)[N+]#N.COC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OC)[N+]#N.Cl.Cl.[Cl-].[Cl-].[Zn+2]
C30H28Cl4N6O6Zn
MFCD00074762
4-Benzamido-2,5-dimethoxybenzenediazonium Salt Zinc Chloride, Fast Blue RR Salt
zinc;N-(4-diazonio-2,5-dimethoxyphenyl)benzenecarboximidate;dichloride;dihydrochloride

Specifications

55663-99-5
C30H28Cl4N6O6Zn
5 g
LLARVKAOQLUCQI-UHFFFAOYSA-L
zinc;N-(4-diazonio-2,5-dimethoxyphenyl)benzenecarboximidate;dichloride;dihydrochloride
134159107
≥95.0% (T)
Azoic Diazo Component 24 (Salt) [for Biochemical Research]
Green-Yellow
MFCD00074762
4-Benzamido-2,5-dimethoxybenzenediazonium Salt Zinc Chloride, Fast Blue RR Salt
COC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OC)[N+]#N.COC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OC)[N+]#N.Cl.Cl.[Cl-].[Cl-].[Zn+2]
775.77
775.77
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-2314

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'