missing translation for 'onlineSavingsMsg'
Learn More

Azelastine Hydrochloride 98.0+%, TCI America™

$68.97 - $348.57

Chemical Identifiers

CAS 79307-93-0
Molecular Formula C22H25Cl2N3O
Molecular Weight (g/mol) 418.36
MDL Number MFCD00242783
InChI Key YEJAJYAHJQIWNU-UHFFFAOYNA-N
Synonym 4-(4-Chlorobenzyl)-2-(1-methylazepan-4-yl)-1-phthalazinone Hydrochloride
PubChem CID 50986914
ChEBI CHEBI:2951
IUPAC Name hydrogen 4-[(4-chlorophenyl)methyl]-2-(1-methylazepan-4-yl)-1,2-dihydrophthalazin-1-one chloride
SMILES [H+].[Cl-].CN1CCCC(CC1)N1N=C(CC2=CC=C(Cl)C=C2)C2=CC=CC=C2C1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A2340100MG
View Documents
TCI America
A2340100MG
100 mg
Each for $68.97
Only null left
 
A23401G
View Documents
TCI America
A23401G
1 g
Each for $348.57
Only null left
 
Description

Description

Specifications

Chemical Identifiers

79307-93-0
418.36
YEJAJYAHJQIWNU-UHFFFAOYNA-N
50986914
hydrogen 4-[(4-chlorophenyl)methyl]-2-(1-methylazepan-4-yl)-1,2-dihydrophthalazin-1-one chloride
C22H25Cl2N3O
MFCD00242783
4-(4-Chlorobenzyl)-2-(1-methylazepan-4-yl)-1-phthalazinone Hydrochloride
CHEBI:2951
[H+].[Cl-].CN1CCCC(CC1)N1N=C(CC2=CC=C(Cl)C=C2)C2=CC=CC=C2C1=O

Specifications

79307-93-0
White
MFCD00242783
4-(4-Chlorobenzyl)-2-(1-methylazepan-4-yl)-1-phthalazinone Hydrochloride
[H+].[Cl-].CN1CCCC(CC1)N1N=C(CC2=CC=C(Cl)C=C2)C2=CC=CC=C2C1=O
418.36
CHEBI:2951
≥98.0% (HPLC,T)
Azelastine Hydrochloride
226°C
C22H25Cl2N3O
100 mg
YEJAJYAHJQIWNU-UHFFFAOYNA-N
hydrogen 4-[(4-chlorophenyl)methyl]-2-(1-methylazepan-4-yl)-1,2-dihydrophthalazin-1-one chloride
50986914
418.36
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : TH9203900

TSCA : No

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'