missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Single Well Medium Profile Reagent Reservoirs
Manufactured with virgin polypropylene for excellent chemical resistance
$846.53 - $891.59
Specifications
Certifications/Compliance | ANSI/SLAS, RNase/DNase-free, non-pyrogenic |
---|---|
Color | Clear |
Capacity (Metric) | 170 mL |
Material | Polypropylene |
Type | Reagent Reservoir |
Catalog Number | Mfr. No. | Sterility | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sterility | Price | Quantity | |||||
14-222-415
|
Axygen™
RESSW384FX |
Non-sterile |
Case of 25 Each for $846.53
|
|
|||||
14-222-416
|
Axygen™
RESSW384FXSI |
Sterile |
Case of 25 Each for $891.59
|
|
|||||
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifications
ANSI/SLAS, RNase/DNase-free, non-pyrogenic | |
170 mL | |
Reagent Reservoir | |
Single-use |
Clear | |
Polypropylene | |
384 |