Learn More
Axygen™ 96-Well Half-Skirt PCR Microplates
Half-skirt amplification plate with single notch
$167.14 - $179.72
Specifications
Certifications/Compliance RNase, DNase-free
Format Standard
Array 8 x 12
Material Polypropylene
No. of Wells 96
View More Specs
Products 3
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
14-223-338
View Documents
Axygen™
PCR96M2HSR
Red
Pack of 10 Each for $168.76
Case of 5 Pack for $843.80
Only null left
14-223-337
View Documents
Axygen™
PCR96M2HSO
Orange
Pack of 10 Each for $179.72
Case of 5 Pack for $898.60
Only null left
14-223-341
View Documents
Axygen™
PCR96M2HSY
Yellow
Pack of 10 Each for $167.14
Case of 5 Pack for $835.70
Only null left
Description
Description
PCR™ products specifically designed for use within PCR thermal transfer and real time PCR applications. A variety of colors are offered to help organize multi-step PCR processes.
  • 96-well PCR half-skirt amplification plate with a single notch
  • Working volume is 300μL
  • Available in a variety of colors
  • Sold 10 plates/unit; 5 units/Cs.
Specifications
Specifications
RNase, DNase-free
8 x 12
96
Round
50/Cs.
Standard
Polypropylene
Non-sterile
PCR thermal transfer, RT-PCR applications
Clear
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'