missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ AX200 ESD Uncoated Gloves

Electrostatic dissipative 18-gauge nylon, uncoated seamless knit glove

$450.26 - $450.26

Specifications

Accelerator Free Yes
Applications Industrial
Coating Material Uncoated
Color Light Gray
Disposable Reusable
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
18-139-752
View Documents
SHOWA™
AX200L08
Small
Case of 72 Pair for $450.26
Only null left
 
18-139-751
View Documents
SHOWA™
AX200M07
Medium
Case of 72 Pair for $450.26
Only null left
 
18-139-750
View Documents
SHOWA™
AX200S06
Large
Case of 72 Pair for $450.26
Only null left
 
18-139-761
View Documents
SHOWA™
AX200XL09
X-Large
Case of 72 Pair for $450.26
Only null left
 
Description

Description

The new SHOWA AX200 has an 18-gauge seamless knit, with nylon static dissipative yarn. It is free of natural rubber latex and is touchscreen compatible, silicone-free and fingerprint-free. Certified under the standard EN 16350, the AX200 by SHOWA meets the requirements for electrostatic properties (ESD). This certification ensures the glove’s safe usage in ATEX zones (working area with risk of explosion and/or high flammability).
Specifications

Specifications

Yes
Uncoated
Reusable
Ambidextrous
Nylon
ESD Gloves
18 gauge
Industrial
Light Gray
No Texture
Yes
Powder Free
Non-sterile
0.8 mm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'