Learn More
Thermo Scientific™ Automation Tips for Qiagen™ Workstations
Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.
Supplier:  Thermo Scientific™ 951011
 View more versions of this product
Catalog No. 02-707-536
Only null left
Explore available promotions
Description
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Qiagen Rosys, Colibri and BioRobot 3000/8000/9600/9604.
  • Available in nonsterile, sterile and with the ART™ filter technology
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)
Specifications
Specifications
Liquid Sensing Tips, 250μL
Filtered
3-10%
Automation
Single-use
250 μL
Sterile
3-10%
96 Tips/Tray, 10 Trays/Case
Sterile and PCR Ready
Product Suggestions
Product Suggestions